Learn More
Cayman Chemical -nbIvolol-d4hydrochlorI 1mg
SDP
Supplier:  Cayman Chemical 266591 MG
Encompass_Preferred
An internal standard for the quantification of nebivolol by GC- or LC-MS
Catalog No. 50-293-4830