Learn More
Aviva Systems Biology Corp NBL1 Antibody - middle region (ARP56637_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56637_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NBL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NBL1. Alternate names: D1S1733E, DAN, DAND1, MGC8972, NB, NO3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61934