Learn More
Aviva Systems Biology Corp NBLA00301 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP42774_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NBLA00301. It was validated on Western Blot. . Alternate names: DEIN, FLJ41674, NBLA00301ApplicationWB. Species reactivity: Human
Catalog No. 50-285-024