Learn More
Selleck Chemical LLC NBMPR S0838-25MG
SDP
Catalog No. 5000042599
Encompass_Preferred
NBMPR (S-(4-Nitrobenzyl)-6-thioinosine) is an inhibitor of ENT1 transporter that crosses the blood-brain barrier and is able to potentiate the neuroprotective effects of adenosine
Catalog No. 50-000-42599 Supplier Selleck Chemical LLC Supplier No. S083825MG
NBMPR (S-(4-Nitrobenzyl)-6-thioinosine) is an inhibitor of ENT1 transporter that crosses the blood-brain barrier and is able to potentiate the neuroprotective effects of adenosine