Learn More
TARGETMOL CHEMICALS INC NBQX 10MG
SDP
Catalog No. 502579958
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NBQX (FG9202) is a potent selective and competitive AMPA receptor antagonist. Neuroprotective and anticonvulsant; active in vivo. purity: 99%
Catalog No. 50-257-9958 Supplier TARGETMOL CHEMICALS INC Supplier No. T411310MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NBQX (FG9202) is a potent selective and competitive AMPA receptor antagonist. Neuroprotective and anticonvulsant; active in vivo. purity: 99%