Learn More
Apexbio Technology LLC NBQX 118876-58-7 10mg
SDP
Catalog No. 5000090855
Encompass_Preferred
NBQX (CAS 118876-58-7) is a potent and selective antagonist of AMPA-type ionotropic glutamate receptors By inhibiting AMPA-mediated excitatory synaptic transmission NBQX disrupts glutamate-induced neuronal signaling thereby attenuating neuronal injury associated with excessive neurotransmitter release This compound has been widely utilized in preclinical studies to investigate neuroprotective and anticonvulsant mechanisms as well as to model and analyze the pathophysiology of neurodegenerative diseases epilepsy and other central nervous system disorders
Catalog No. 50-000-90855 Supplier Apexbio Technology LLC Supplier No. B628010
NBQX (CAS 118876-58-7) is a potent and selective antagonist of AMPA-type ionotropic glutamate receptors By inhibiting AMPA-mediated excitatory synaptic transmission NBQX disrupts glutamate-induced neuronal signaling thereby attenuating neuronal injury associated with excessive neurotransmitter release This compound has been widely utilized in preclinical studies to investigate neuroprotective and anticonvulsant mechanisms as well as to model and analyze the pathophysiology of neurodegenerative diseases epilepsy and other central nervous system disorders