Learn More
Apexbio Technology LLC NBQX 118876-58-7 50mg
SDP
Catalog No. 5000089801
Encompass_Preferred
NBQX (CAS 118876-58-7) is a potent and selective antagonist of AMPA-type ionotropic glutamate receptors By inhibiting AMPA-mediated excitatory synaptic transmission NBQX disrupts glutamate-induced neuronal signaling thereby attenuating neuronal injury associated with excessive neurotransmitter release This compound has been widely utilized in preclinical studies to investigate neuroprotective and anticonvulsant mechanisms as well as to model and analyze the pathophysiology of neurodegenerative diseases epilepsy and other central nervous system disorders
Catalog No. 50-000-89801 Supplier Apexbio Technology LLC Supplier No. B628050
NBQX (CAS 118876-58-7) is a potent and selective antagonist of AMPA-type ionotropic glutamate receptors By inhibiting AMPA-mediated excitatory synaptic transmission NBQX disrupts glutamate-induced neuronal signaling thereby attenuating neuronal injury associated with excessive neurotransmitter release This compound has been widely utilized in preclinical studies to investigate neuroprotective and anticonvulsant mechanisms as well as to model and analyze the pathophysiology of neurodegenerative diseases epilepsy and other central nervous system disorders