Learn More
Medchemexpress LLC NBQX | 118876-58-7 | C12H8N4O6S | 25 MG
SDP
Catalog No. 5000375975
Encompass_Preferred
NBQX (FG9202) is a highly selective and competitive AMPA receptor antagonist with neuroprotective and anticonvulsant activity.
  • High affinity for AMPA and kainate binding sites.
  • Minimal or no affinity for the glutamate recognition site on the NMDA receptor complex.
  • Effective in decreasing seizures induced by PTZ.
  • Neuroprotective in a focal ischaemia model.
Catalog No. 50-003-75975 Supplier Medchemexpress LLC Supplier No. 25MGHY15068
NBQX (FG9202) is a highly selective and competitive AMPA receptor antagonist with neuroprotective and anticonvulsant activity.
  • High affinity for AMPA and kainate binding sites.
  • Minimal or no affinity for the glutamate recognition site on the NMDA receptor complex.
  • Effective in decreasing seizures induced by PTZ.
  • Neuroprotective in a focal ischaemia model.