Learn More
Medchemexpress LLC NBQX disodium | 479347-86-9 | C12H6N4Na2O6S | 1 ML
SDP
Catalog No. 5000375114
Encompass_Preferred
NBQX disodium is a highly selective and competitive AMPA receptor antagonist, known for its neuroprotective and anticonvulsant activities. It displays high affinity for AMPA and kainate binding sites.
  • Highly selective and competitive AMPA receptor antagonist
  • Exhibits neuroprotective activity
  • Demonstrates anticonvulsant activity
  • Targets AMPA receptors
  • Decreases seizures induced by PTZ in animal models
  • Neuroprotective in focal ischaemia models
Catalog No. 50-003-75114 Supplier Medchemexpress LLC Supplier No. HY15068A10MM/1ML
NBQX disodium is a highly selective and competitive AMPA receptor antagonist, known for its neuroprotective and anticonvulsant activities. It displays high affinity for AMPA and kainate binding sites.
  • Highly selective and competitive AMPA receptor antagonist
  • Exhibits neuroprotective activity
  • Demonstrates anticonvulsant activity
  • Targets AMPA receptors
  • Decreases seizures induced by PTZ in animal models
  • Neuroprotective in focal ischaemia models