Learn More
Medchemexpress LLC NBQX disodium | 479347-86-9 | C12H6N4Na2O6S | 50 MG
SDP
Catalog No. 5000375092
Encompass_Preferred
NBQX disodium (FG9202 disodium) is a highly selective and competitive AMPA receptor antagonist. It exhibits neuroprotective and anticonvulsant activity. This product shows high affinity for AMPA and kainate binding sites, with minimal or no affinity for the glutamate recognition site on the NMDA receptor complex. It has also demonstrated neuroprotective effects in a focal ischaemia model in rats.
  • Highly selective and competitive AMPA receptor antagonist
  • Possesses neuroprotective activity
  • Exhibits anticonvulsant activity
  • High affinity for AMPA and kainate binding sites
  • Minimal affinity for the glutamate recognition site on the NMDA receptor complex
Catalog No. 50-003-75092 Supplier Medchemexpress LLC Supplier No. HY15068A50MG
NBQX disodium (FG9202 disodium) is a highly selective and competitive AMPA receptor antagonist. It exhibits neuroprotective and anticonvulsant activity. This product shows high affinity for AMPA and kainate binding sites, with minimal or no affinity for the glutamate recognition site on the NMDA receptor complex. It has also demonstrated neuroprotective effects in a focal ischaemia model in rats.
  • Highly selective and competitive AMPA receptor antagonist
  • Possesses neuroprotective activity
  • Exhibits anticonvulsant activity
  • High affinity for AMPA and kainate binding sites
  • Minimal affinity for the glutamate recognition site on the NMDA receptor complex