Learn More
TARGETMOL CHEMICALS INC NBQX DISODIUM 5MG
SDP
Catalog No. 502580736
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. NBQX Disodium (FG9202 disodium) is a highly selective competitive AMPA receptor antagonist with neuroprotective and anticonvulsant activity. NBQX Disodium is a salt of NBQX. NBQX is a potent and selective AMPA/rhodosinate receptor antagonist that can be used to antagonize the excitatory toxicity of glutamate. It has anticonvulsant effects in rodent models. purity: 98%
Catalog No. 50-258-0736 Supplier TARGETMOL CHEMICALS INC Supplier No. T258535MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. NBQX Disodium (FG9202 disodium) is a highly selective competitive AMPA receptor antagonist with neuroprotective and anticonvulsant activity. NBQX Disodium is a salt of NBQX. NBQX is a potent and selective AMPA/rhodosinate receptor antagonist that can be used to antagonize the excitatory toxicity of glutamate. It has anticonvulsant effects in rodent models. purity: 98%