Learn More
Apexbio Technology LLC NBQX disodium salt 479347-86-9 50mg
SDP
Catalog No. 5000091813
Encompass_Preferred
NBQX disodium salt (CAS 479347-86-9) is a selective competitive antagonist of -amino-3-hydroxy-5-methyl-4-isoxazolepropionate (AMPA) receptors a subtype of ionotropic glutamate receptors By inhibiting AMPA receptor-mediated excitatory neurotransmission NBQX disodium salt is utilized to study synaptic plasticity and excitotoxicity mechanisms This compound has been shown to exhibit neuroprotective and anticonvulsant properties in preclinical models and is widely used in research on neurological disorders such as Alzheimer s disease epilepsy and schizophrenia to elucidate the physiological and pathological roles of AMPA receptor signaling
Catalog No. 50-000-91813 Supplier Apexbio Technology LLC Supplier No. B656650
NBQX disodium salt (CAS 479347-86-9) is a selective competitive antagonist of -amino-3-hydroxy-5-methyl-4-isoxazolepropionate (AMPA) receptors a subtype of ionotropic glutamate receptors By inhibiting AMPA receptor-mediated excitatory neurotransmission NBQX disodium salt is utilized to study synaptic plasticity and excitotoxicity mechanisms This compound has been shown to exhibit neuroprotective and anticonvulsant properties in preclinical models and is widely used in research on neurological disorders such as Alzheimer s disease epilepsy and schizophrenia to elucidate the physiological and pathological roles of AMPA receptor signaling