Learn More
Cayman Chemical NBQX (sodium salt), Purity: ≥98%, CAS: 479347-86-9, 50 mg
SDP
Catalog No. 502969479
Encompass_Preferred
A water-soluble receptor antagonist which targets both AMPA- and kainate-sensitive glutamate receptors (IC50 = 0.15 and 4.8 µM, respectively); does not block the NMDA-sensitive glutamate receptor (IC50 = ≥ 90 µM)
Catalog No. 50-296-9479 Supplier Cayman Chemical Supplier No. 1491450 MG
A water-soluble receptor antagonist which targets both AMPA- and kainate-sensitive glutamate receptors (IC50 = 0.15 and 4.8 µM, respectively); does not block the NMDA-sensitive glutamate receptor (IC50 = ≥ 90 µM)