Learn More
Aviva Systems Biology Corp Nbr1 Antibody - C-terminal region (ARP53647_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53647_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nbr1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: mKIAA0049ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-863