Learn More
Electron Microscopy Sciences NBS 25 Chart 20 Lines per mm
SDP
Catalog No. 501901579
Encompass_Preferred
This consists of elements of 5 bars ranging from 1 line per mm to 18 lines per mm, with adjacent numbers indicating the number of lines per mm. Produced as a vacuum deposited chrome image on 75 mm x 75 mm glass.
Catalog No. 50-190-1579 Supplier Electron Microscopy Sciences Supplier No. 6809810
This consists of elements of 5 bars ranging from 1 line per mm to 18 lines per mm, with adjacent numbers indicating the number of lines per mm. Produced as a vacuum deposited chrome image on 75 mm x 75 mm glass.