Learn More
Sigma Aldrich Fine Chemicals Biosciences Corning(R) NBS(TM) 384 well microplates surface treatment, Non-binding Surface, black polystyrene, flat bottom, non-sterile, lid: no, pack of 10 | 50EA
Catalog No. NC1959043
Encompass
Corning(R) NBS(TM) 384 well microplates surface treatment, Non-binding Surface, black polystyrene, flat bottom, non-sterile, lid: no, pack of 10 | 50EA
Catalog No. NC1959043 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS382050EA
Corning(R) NBS(TM) 384 well microplates surface treatment, Non-binding Surface, black polystyrene, flat bottom, non-sterile, lid: no, pack of 10 | 50EA