Learn More
Electron Microscopy Sciences NBS 5 Bar Test Ref 1963A BS 4657
SDP
Catalog No. 501901580
Encompass_Preferred
This consists of elements of 5 bars ranging from 1 line per mm to 18 lines per mm, with adjacent numbers indicating the number of lines per mm. Produced as a vacuum deposited chrome image on B270 75 mm x 75 mm glass.
Catalog No. 50-190-1580 Supplier Electron Microscopy Sciences Supplier No. 6809811
This consists of elements of 5 bars ranging from 1 line per mm to 18 lines per mm, with adjacent numbers indicating the number of lines per mm. Produced as a vacuum deposited chrome image on B270 75 mm x 75 mm glass.