Learn More
Biotium NBT (Nitro Blue Tetrazolium, Chloride), 5 g
Catalog No. 501964096
Encompass_Preferred
NBT (Nitro blue tetrazolium, chloride) is an oxidant and is widely used in conjunction with BCIP or BCIP derivatives to detect phosphatase activity. NBT is also a substrate for dehydrogenate and other oxidizes. Thus, it is often used to assess cell viability, proliferation and cytotoxicity, and is also frequently used in histochemistry studies. The colorless NBT is soluble in water, but forms a dark blue-colored, highly insoluble precipitate after being enzymatically reduced. Properties: White solid soluble in water and DMSO; Store at 4°C; C40H30Cl2N10O6; MW: 817.7; [298-83-9]
Catalog No. 50-196-4096 Supplier Biotium Supplier No. 100082
NBT (Nitro blue tetrazolium, chloride) is an oxidant and is widely used in conjunction with BCIP or BCIP derivatives to detect phosphatase activity. NBT is also a substrate for dehydrogenate and other oxidizes. Thus, it is often used to assess cell viability, proliferation and cytotoxicity, and is also frequently used in histochemistry studies. The colorless NBT is soluble in water, but forms a dark blue-colored, highly insoluble precipitate after being enzymatically reduced. Properties: White solid soluble in water and DMSO; Store at 4°C; C40H30Cl2N10O6; MW: 817.7; [298-83-9]