Learn More
Alkali Scientific NBT [Nitro Blue Tetrazolium] [2,2' Di P Nitrophenyl 5, 5' Diphenyl 3, 3' (3,3' Dimethoxy 4, 4' Diphenylene) Ditetrazolium Chloride], 500 Milligrams
SDP
Catalog No. 5000661419
Encompass_Preferred
Nitro blue tetrazolium (NBT) is a chromogenic reagent used in enzyme detection and histochemical applications. When reduced by enzymes such as dehydrogenases or reductases, NBT produces a blue formazan precipitate, making it useful for detecting enzyme activity in biological samples. The 500mg quantity is ideal for preparing multiple assays or for large-scale enzyme activity screenings. NBT is commonly used in immunohistochemistry, immunoblotting, and various biochemical assays for detecting enzyme activity, metabolic processes, and cell viability. This reagent is essential in a variety of laboratory applications, offering a simple and sensitive method for enzyme detection and analysis.
Catalog No. 50-006-61419 Supplier Alkali Scientific Supplier No. NBT004CP
Nitro blue tetrazolium (NBT) is a chromogenic reagent used in enzyme detection and histochemical applications. When reduced by enzymes such as dehydrogenases or reductases, NBT produces a blue formazan precipitate, making it useful for detecting enzyme activity in biological samples. The 500mg quantity is ideal for preparing multiple assays or for large-scale enzyme activity screenings. NBT is commonly used in immunohistochemistry, immunoblotting, and various biochemical assays for detecting enzyme activity, metabolic processes, and cell viability. This reagent is essential in a variety of laboratory applications, offering a simple and sensitive method for enzyme detection and analysis.