Learn More
Cayman Chemical N-butyryl-L-Homocysteine thiolactone, Purity: ≥95%, CAS: 202284-85-3, 25 mg
SDP
Catalog No. 502888822
Encompass_Preferred
An analog of N-butyryl-L-HSL, the small diffusible signaling molecule involved in quorum sensing, controlling gene expression, and cellular metabolism; induces violacein expression in C. violaceum mutants usually not able to produce AHLs
Catalog No. 50-288-8822 Supplier Cayman Chemical Supplier No. 1001120425 MG
An analog of N-butyryl-L-HSL, the small diffusible signaling molecule involved in quorum sensing, controlling gene expression, and cellular metabolism; induces violacein expression in C. violaceum mutants usually not able to produce AHLs