Learn More
Cayman Chemical NbutyrylLHomocysteine thiolac
SDP
Catalog No. 502889345
Encompass_Preferred
An analog of N-butyryl-L-HSL, the small diffusible signaling molecule involved in quorum sensing, controlling gene expression, and cellular metabolism; induces violacein expression in C. violaceum mutants usually not able to produce AHLs
Catalog No. 50-288-9345 Supplier Cayman Chemical Supplier No. 1001120410 MG
An analog of N-butyryl-L-HSL, the small diffusible signaling molecule involved in quorum sensing, controlling gene expression, and cellular metabolism; induces violacein expression in C. violaceum mutants usually not able to produce AHLs