Learn More
Cayman Chemical NbutyrylLHomoserine lactone
Supplier: Cayman Chemical 1000789825 MG
A bacterial quorum sensing signal molecule; regulates virulence in general, infection prevention, and formation of biofilms