Learn More
Cayman Chemical NbutyrylLHomoserine lactone
SDP
Catalog No. 502888612
Encompass_Preferred
A bacterial quorum sensing signal molecule; regulates virulence in general, infection prevention, and formation of biofilms
Catalog No. 50-288-8612 Supplier Cayman Chemical Supplier No. 1000789850 MG
A bacterial quorum sensing signal molecule; regulates virulence in general, infection prevention, and formation of biofilms