Learn More
Cayman Chemical NbutyrylLHomoserine lactone
SDP
Catalog No. 502891170
Encompass_Preferred
A bacterial quorum sensing signal molecule; regulates virulence in general, infection prevention, and formation of biofilms
Catalog No. 50-289-1170 Supplier Cayman Chemical Supplier No. 100078985 MG
A bacterial quorum sensing signal molecule; regulates virulence in general, infection prevention, and formation of biofilms