Learn More
Crescent Chemical Co Inc NC 2 NITROCELLULOSE MEMBRANE
SDP
Catalog No. 50249214
Encompass_Preferred
NC 2 Nitrocellulose Membrane, white Pore size 0.2 µm, format: 20 x 20 cm Especially for use with proteins of low molecular weight (\Nitrocellulose membranes are the most popular membranes for Western, Southern and Northern Blotting. The membranes bind both proteins and nucleic acids.
Catalog No. 50-249-214 Supplier Crescent Chemical Co Inc Supplier No. SE71223.01
NC 2 Nitrocellulose Membrane, white Pore size 0.2 µm, format: 20 x 20 cm Especially for use with proteins of low molecular weight (\Nitrocellulose membranes are the most popular membranes for Western, Southern and Northern Blotting. The membranes bind both proteins and nucleic acids.