Learn More
Crescent Chemical Co Inc NC 2 NITROCELLULOSE MEMBRANE
SDP
Supplier:  Crescent Chemical Co Inc SE71224.01
Encompass_Preferred
NC 2 Nitrocellulose Membrane, white Pore size 0.2 µm, format: 30 cm x 3 m Especially for use with proteins of low molecular weight (\Nitrocellulose membranes are the most popular membranes for Western, Southern and Northern Blotting. The membranes bind both proteins and nucleic acids
Catalog No. 50-249-215
 Restricted Item