Learn More
CHEMOMETEC NC-200™ PQ Test Kit 4
SDP
Catalog No. NC1648096
Encompass
The NC-200™ PQ Test Kit 4 is designed to validate the functionality of NucleoCounter NC-200™ instrument. Compared to the standard PQ Test Kit, PQ Test Kit 4 offers an extended range of bead concentrations and additional viability controls. Performance qualification (PQ) of laboratory equipment is an important part of the validation of the instrument. Its purpose is to verify and document that the instrument is working reproducibly within the specified working range and limits. The NC-200™ PQ Test Kit 4 consists of three preparations in different concentrations of fluorescent beads with different blue/green ratios and a PQ test plan with information on the tests to be performed on the instrument.
Catalog No. NC1648096 Supplier CHEMOMETEC Supplier No. 9120013
The NC-200™ PQ Test Kit 4 is designed to validate the functionality of NucleoCounter NC-200™ instrument. Compared to the standard PQ Test Kit, PQ Test Kit 4 offers an extended range of bead concentrations and additional viability controls. Performance qualification (PQ) of laboratory equipment is an important part of the validation of the instrument. Its purpose is to verify and document that the instrument is working reproducibly within the specified working range and limits. The NC-200™ PQ Test Kit 4 consists of three preparations in different concentrations of fluorescent beads with different blue/green ratios and a PQ test plan with information on the tests to be performed on the instrument.