Learn More
Cayman Chemical nC 55-0396 10mg
SDP
Catalog No. 502979970
Encompass_Preferred
A selective inhibitor of T-type calcium channels (IC50 = 6.8 μM); exhibits no activity against L-type calcium channels up to 100 μM; inhibits tremor in mice (20 mg/kg and 12.5 mg/kg in GABAA α1-null and harmaline-induced tremor models, respectively); inhibits Kv channels (IC50 = 80 nM)
Catalog No. 50-297-9970 Supplier Cayman Chemical Supplier No. 1721610 MG
A selective inhibitor of T-type calcium channels (IC50 = 6.8 μM); exhibits no activity against L-type calcium channels up to 100 μM; inhibits tremor in mice (20 mg/kg and 12.5 mg/kg in GABAA α1-null and harmaline-induced tremor models, respectively); inhibits Kv channels (IC50 = 80 nM)