Learn More
Cayman Chemical NNC-711 (hydrochloride), Purity: ≥98%, CAS: 145645-62-1, 10 mg
SDP
Catalog No. 502990523
Encompass_Preferred
A GAT-1 inhibitor (IC50 = 0.04 µM for the human transporter); selective for GAT-1 over human GAT-3 and BGT-3 (IC50s = 1,700 and 622 µM, respectively), rat GAT-2 (IC50 = 171 µM), and a panel of 21 receptors and ion channels (IC50s = >50 µM for all) in radioligand binding assays; inhibits GABA uptake in rat synaptosome preparations, neurons, and glia (IC50s = 47, 1,238, and 636 nM, respectively); inhibits PTZ- or DMCM-induced clonic seizures in mice (ED50s = 1.2 and 3 mg/kg, respectively), as well as audiogenic tonic seizures in mice (ED50 = 0.23 mg/kg) and PTZ-induced tonic seizures in rats (ED50 = 1.7 mg/kg); intrathecal administration decreases mechanical allodynia and thermal hyperalgesia in a rat model of CCI-induced neuropathic pain at 100 or 200 µg/animal
Catalog No. 50-299-0523 Supplier Cayman Chemical Supplier No. 2968710 MG
A GAT-1 inhibitor (IC50 = 0.04 µM for the human transporter); selective for GAT-1 over human GAT-3 and BGT-3 (IC50s = 1,700 and 622 µM, respectively), rat GAT-2 (IC50 = 171 µM), and a panel of 21 receptors and ion channels (IC50s = >50 µM for all) in radioligand binding assays; inhibits GABA uptake in rat synaptosome preparations, neurons, and glia (IC50s = 47, 1,238, and 636 nM, respectively); inhibits PTZ- or DMCM-induced clonic seizures in mice (ED50s = 1.2 and 3 mg/kg, respectively), as well as audiogenic tonic seizures in mice (ED50 = 0.23 mg/kg) and PTZ-induced tonic seizures in rats (ED50 = 1.7 mg/kg); intrathecal administration decreases mechanical allodynia and thermal hyperalgesia in a rat model of CCI-induced neuropathic pain at 100 or 200 µg/animal