Learn More
CHEMOMETEC Via2 Cassette
SDP
Catalog No. NC2167110
Encompass
The Via2-Cassette is a single-use cassette for preparing blood samples for the NucleoCounter NC-202 system. It enables automated cell counting and viability measurements. The cassette contains all necessary reagents. It has two chambers - one for cell lysis and one for DNA staining. The cassette is inserted into the NC-202 system where image cytometry is used to count and analyze cells after preparation. The cassette streamlines workflow and minimizes hands-on time. It is stable at room temperature and has a 12 month shelf life. The cassette works with all common anticoagulants and can process samples from whole blood, bone marrow, CSF, and cell cultures.
This item is not returnable. View return policy
Catalog No. NC2167110 Supplier CHEMOMETEC Supplier No. 9410024
The Via2-Cassette is a single-use cassette for preparing blood samples for the NucleoCounter NC-202 system. It enables automated cell counting and viability measurements. The cassette contains all necessary reagents. It has two chambers - one for cell lysis and one for DNA staining. The cassette is inserted into the NC-202 system where image cytometry is used to count and analyze cells after preparation. The cassette streamlines workflow and minimizes hands-on time. It is stable at room temperature and has a 12 month shelf life. The cassette works with all common anticoagulants and can process samples from whole blood, bone marrow, CSF, and cell cultures.