Learn More
Cayman Chemical Inecalcitol, Purity: ≥98%, CAS: 163217-09-2, 10 mg
SDP
Catalog No. 502917571
Encompass_Preferred
A vitamin D receptor agonist; induces reporter gene expression in VDR-expressing COS-1 cells at 1 nM; induces differentiation of HL-60 and MG-63 cells (EC50s = 6.2 and 0.18 nM, respectively); reduces the proliferation of MCF-7 cells (EC50 = 4.1 nM); protects primary human keratinocytes against DNA damage induced by UVB irradiation; inhibits acetylcholine- or ATP-induced contractions of aortic rings isolated from spontaneously hypertensive rats at 100 nM; unlike calcitriol, it does not increase serum calcium levels or decrease tibia calcium levels in mice at 80 µg/kg
Catalog No. 50-291-7571 Supplier Cayman Chemical Supplier No. 3728810 MG
A vitamin D receptor agonist; induces reporter gene expression in VDR-expressing COS-1 cells at 1 nM; induces differentiation of HL-60 and MG-63 cells (EC50s = 6.2 and 0.18 nM, respectively); reduces the proliferation of MCF-7 cells (EC50 = 4.1 nM); protects primary human keratinocytes against DNA damage induced by UVB irradiation; inhibits acetylcholine- or ATP-induced contractions of aortic rings isolated from spontaneously hypertensive rats at 100 nM; unlike calcitriol, it does not increase serum calcium levels or decrease tibia calcium levels in mice at 80 µg/kg