Learn More
Aviva Systems Biology Corp NCAM1 Antibody - C-terminal region (ARP59939_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59939_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NCAM1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human NCAM1. Alternate names: CD56, MSK39, NCAMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-883