Learn More
Sino Biological NCAM1 / CD56 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10673T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NCAM1 / CD56 (rh NCAM1 / CD56; Catalog#10673-H08H; NP_001070150.1; Met1-Pro603). NCAM1 / CD56 specific IgG was purified by Human NCAM1 / CD56 affinity chromatography.
Catalog No. 50-224-8539