Learn More
Sino Biological NCAM1/CD56 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 108577T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse NCAM1/CD56, and purified by antigen affinity chromatography.
Catalog No. 50-224-8130