Learn More
Cayman Chemical NCAM1CD56CTerm RabbIt Mo 100uL
SDP
Catalog No. 502949343
Encompass_Preferred
A catechol estrogen and minor metabolite of estrogen production is increased in microsomes prepared from human mammary adenocarcinoma and fibroadenoma microsomes compared with microsomes prepared from non-cancerous tissues induces renal tumor formation in male hamsters at 25 mg/animal
Catalog No. 50-294-9343 Supplier Cayman Chemical Supplier No. 32257100 UL
A catechol estrogen and minor metabolite of estrogen production is increased in microsomes prepared from human mammary adenocarcinoma and fibroadenoma microsomes compared with microsomes prepared from non-cancerous tissues induces renal tumor formation in male hamsters at 25 mg/animal