Learn More
Aviva Systems Biology Corp NCAPD2 Antibody - C-terminal region (ARP46210_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46210_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NCAPD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NCAPD2. Alternate names: CAP-D2, CNAP1, KIAA0159, hCAP-D2ApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-59269