Learn More
Aviva Systems Biology Corp NCAPH Antibody - C-terminal region (ARP46227_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46227_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NCAPH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NCAPH. Alternate names: BRRN1, CAP-H, HCAP-HApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59278