Learn More
Aviva Systems Biology Corp NCAPH2 Antibody - N-terminal region (ARP55483_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55483_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NCAPH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NCAPH2. Alternate names: 384D8-2, 384D8_6, CAP-H2, MGC15858, MGC18000, MGC2455, MGC4133, MGC5305, MGC8640, hCAP-H2, CAPH2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61520