Learn More
TARGETMOL CHEMICALS INC NCB-0846 10MG
SDP
Catalog No. 502576831
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NCB-0846 is an orally active TNIK inhibitor (IC50 21 nM). purity: 99%
Catalog No. 50-257-6831 Supplier TARGETMOL CHEMICALS INC Supplier No. T401110MG
Add to Cart
Add to Cart
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NCB-0846 is an orally active TNIK inhibitor (IC50 21 nM). purity: 99%