Learn More
Cayman Chemical N-Cbz-Gly-Pro-Gly-Gly-Pro-Ala-OH (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502950831
Encompass_Preferred
A peptide collagenase substrate; has been used to characterize the collagenolytic activity of bacterial collagenases in cell-free assays
Catalog No. 50-295-0831 Supplier Cayman Chemical Supplier No. 3334310 MG
A peptide collagenase substrate; has been used to characterize the collagenolytic activity of bacterial collagenases in cell-free assays