Learn More
Cayman Chemical N-Cbz-Gly-Pro-Gly-Gly-Pro-Ala-OH (trifluoroacetate salt), Purity: ≥98%, 50 mg
SDP
Catalog No. 502950503
Encompass_Preferred
A peptide collagenase substrate; has been used to characterize the collagenolytic activity of bacterial collagenases in cell-free assays
Catalog No. 50-295-0503 Supplier Cayman Chemical Supplier No. 3334350 MG
A peptide collagenase substrate; has been used to characterize the collagenolytic activity of bacterial collagenases in cell-free assays