Learn More
Medchemexpress LLC NCC-149 | 1316652-41-1 | 98.7% | 326.37 g/mol | C16H14N4O2S | 1 ML
SDP
Catalog No. 5000228570
Encompass_Preferred
NCC-149 is a selective HDAC8 inhibitor intended for in vitro research applications, including neural differentiation studies. It is supplied as either a 10 mM solution in DMSO or as a solid for reconstitution, and is characterized by reported high purity and a defined molecular weight.
  • Selective HDAC8 inhibition suitable for mechanistic and cellular studies.
  • Useful for neural differentiation research and related assays.
  • Available as 10 mM solution in DMSO or as solid for preparation of stock solutions.
  • High reported purity supports reproducible experimental results.
  • Defined molecular weight and formula simplify calculation of working concentrations.
Catalog No. 50-002-28570 Supplier Medchemexpress LLC Supplier No. HY11734810MM 1ML
NCC-149 is a selective HDAC8 inhibitor intended for in vitro research applications, including neural differentiation studies. It is supplied as either a 10 mM solution in DMSO or as a solid for reconstitution, and is characterized by reported high purity and a defined molecular weight.
  • Selective HDAC8 inhibition suitable for mechanistic and cellular studies.
  • Useful for neural differentiation research and related assays.
  • Available as 10 mM solution in DMSO or as solid for preparation of stock solutions.
  • High reported purity supports reproducible experimental results.
  • Defined molecular weight and formula simplify calculation of working concentrations.