Learn More
Aviva Systems Biology Corp NCCRP1 Antibody - N-terminal region (ARP54371_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54371_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC342897. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LOC342897. Alternate names: FBXO50, NCCRP-1ApplicationWB. Species reactivity: Human
Catalog No. 50-285-915