Learn More
Aviva Systems Biology Corp NCDN Antibody - N-terminal region (ARP54995_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54995_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NCDN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NCDN. Alternate names: KIAA0607ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61390