Learn More
Sino Biological NCF2 / NCF-2 / P67phox Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 15669RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NCF2 / NCF-2 / P67phox (rh NCF2; Catalog#15669-H20B; AAH01606.1; Met1-Val526). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-7980