Learn More
Aviva Systems Biology Corp NCF4 Antibody - middle region (ARP46010_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46010_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NCF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NCF4. Alternate names: MGC3810, NCF, P40PHOX, SH3PXD4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59176