Learn More
Medchemexpress LLC Ncgc 607 | 1462267-07-7 | 99.5% | 5 MG
SDP
Catalog No. 5000234330
Encompass_Preferred
NCGC 607 is a small-molecule, noninhibitory chaperone of glucocerebrosidase (GCase) used for research applications; it has been reported to increase GCase activity and reduce α-synuclein, making it relevant to studies of Gaucher disease and synucleinopathies.
  • Noninhibitory glucocerebrosidase chaperone.
  • Increases GCase activity and protein levels.
  • Reduces α-synuclein and glycolipid accumulation.
  • High purity (99.53%) suitable for research assays.
  • Small-molecule, solid form; soluble in DMSO for biochemical use.
Catalog No. 50-002-34330 Supplier Medchemexpress LLC Supplier No. HY1200345MG
NCGC 607 is a small-molecule, noninhibitory chaperone of glucocerebrosidase (GCase) used for research applications; it has been reported to increase GCase activity and reduce α-synuclein, making it relevant to studies of Gaucher disease and synucleinopathies.
  • Noninhibitory glucocerebrosidase chaperone.
  • Increases GCase activity and protein levels.
  • Reduces α-synuclein and glycolipid accumulation.
  • High purity (99.53%) suitable for research assays.
  • Small-molecule, solid form; soluble in DMSO for biochemical use.