Learn More
Medchemexpress LLC HY-114043 5mg Medchemexpress, NCGC00092410 CAS:442898-34-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1140435MG
Encompass_Preferred
Medchemexpress, HY-114043 5mg NCGC00092410 CAS:442898-34-2 NCGC00092410 is a potent, selective, and nonsugar glucocerebrosidase (GC) inhibitor, with an IC50 of 31 nM. NCGC00092410 shows no activity against the related hydrolases at concentrations up to 77 μM. NCGC00092410, a GC chaperone, and increases the activity and lysosomal localization of glucocerebrosidase in mutant cell lines. NCGC00092410 can be used for the research of Gaucher disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0866