Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ncgc00138783 TFA | 00-00-0 | 99.4% | 617.62 g·mol⁻¹ | C28H27F4N7O3S | 5 MG
SDP

Catalog No. 5000222677
Encompass_Preferred

NCGC00138783 TFA is a small-molecule inhibitor that selectively targets the CD47/SIRPα immune checkpoint, blocking the CD47-SIRPα interaction with reported in vitro activity (IC50 ≈ 50 μM). Supplied as the trifluoroacetic acid (TFA) salt for research applications.

  • Selective inhibitor of the CD47/SIRPα immune checkpoint.
  • Reported IC50 approximately 50 μM in biochemical assays.
  • Supplied as the trifluoroacetic acid (TFA) salt.
  • High purity (99.4%).
  • Molecular weight 617.62 g·mol⁻¹.
  • Store at 4 °C; protect from moisture and light.

Catalog No. 50-002-22677 Supplier Medchemexpress LLC Supplier No. HY143224B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NCGC00138783 TFA is a small-molecule inhibitor that selectively targets the CD47/SIRPα immune checkpoint, blocking the CD47-SIRPα interaction with reported in vitro activity (IC50 ≈ 50 μM). Supplied as the trifluoroacetic acid (TFA) salt for research applications.

  • Selective inhibitor of the CD47/SIRPα immune checkpoint.
  • Reported IC50 approximately 50 μM in biochemical assays.
  • Supplied as the trifluoroacetic acid (TFA) salt.
  • High purity (99.4%).
  • Molecular weight 617.62 g·mol⁻¹.
  • Store at 4 °C; protect from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.