Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ncgc00138783 tfa | 896700-07-5 | 99.4% | C28H27F4N7O3S | 1 MG
SDP

Catalog No. 5000356145
Encompass_Preferred

Ncgc00138783 TFA is a selective inhibitor targeting the CD47/SIRPα axis, with an IC50 of 50 μM. It directly blocks the interaction between the CD47 and SIRPα axis.

  • Target: CD47/SIRPα axis
  • Selective inhibitor
  • Blocks CD47/SIRPα interaction
  • Molecular weight: 617.62
  • Appearance: Solid, white to light yellow
  • Purity: 99.4%
  • Applications in immunology/inflammation

Catalog No. 50-003-56145 Supplier Medchemexpress LLC Supplier No. HY143224B1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ncgc00138783 TFA is a selective inhibitor targeting the CD47/SIRPα axis, with an IC50 of 50 μM. It directly blocks the interaction between the CD47 and SIRPα axis.

  • Target: CD47/SIRPα axis
  • Selective inhibitor
  • Blocks CD47/SIRPα interaction
  • Molecular weight: 617.62
  • Appearance: Solid, white to light yellow
  • Purity: 99.4%
  • Applications in immunology/inflammation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.